Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1s Antibody (006), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289344B
Description
Complement Component C1s Monoclonal antibody specifically detects Complement Component C1s in Human samples. It is validated for ELISASpecifications
Complement Component C1s | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
716 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
006 | |
ELISA | |
basic proline-rich peptide IB-1, C1 esterase, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C1s (NP_001725.1; Met 1-Asp 688). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction