Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C1s Antibody (9), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105023AF405
Description
Complement Component C1s Monoclonal specifically detects Complement Component C1s in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Complement Component C1s | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C1S | |
C1s proenzyme purified from human plasma | |
0.1 mL | |
Primary | |
Specific for C1s proenzyme and activated free C1s | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry | |
9 | |
ELISA, Immunohistochemistry | |
basic proline-rich peptide IB-1, C1 esterase, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
Mouse | |
Protein A purified | |
RUO | |
716 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction