Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C2 Antibody (014), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289301APC
Description
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for ELISASpecifications
| Complement Component C2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 717 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 014 | |
| Sandwich ELISA Capture | |
| C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C2 (Met 1-Leu 752; Accession#: NP_000054.2). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction