Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C2 Antibody (014), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289301G
Description
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for ELISA, ELISASpecifications
Complement Component C2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
717 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
014 | |
ELISA, Sandwich ELISA | |
C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C2 (NP_000054.2; Met1-Leu752). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction