Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C3aR Antibody (3G7), Janelia Fluor™ 646 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252674JF646
Description
Complement Component C3aR Monoclonal specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence.Specifications
Complement Component C3aR | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C3AR1 | |
The 2e loop of the C3a receptor. | |
0.1 mL | |
719 | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
3G7 | |
Flow Cytometry, ELISA, Immunofluorescence | |
AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
Rabbit | |
Protein A or G purified | |
Primary | |
Binds specifically to the 2e loop of the human C3a receptor, residues 185-193. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction