Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C3aR Antibody (3G7), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252674MFV500
Description
Complement Component C3aR Monoclonal antibody specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, CyTOFSpecifications
| Complement Component C3aR | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence, CyTOF | |
| 3G7 | |
| Flow Cytometry, ELISA, Immunofluorescence, CyTOF-ready | |
| AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
| The 2e loop of the C3a receptor. | |
| 0.1 mL | |
| GPCR | |
| 719 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction