Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C3aR Antibody (620613) [DyLight 755], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB10417Z
Description
Complement Component C3aR Monoclonal antibody specifically detects Complement Component C3aR in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Complement Component C3aR | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
620613 | |
Flow Cytometry, CyTOF-ready | |
AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
Chinese hamster ovary cell line CHO-derived transfected with mouse C3aR, Accession # O09047 | |
0.1 mL | |
GPCR | |
719 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction