Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C3aR Antibody (HC3aRZ8), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314618AF488
This item is not returnable.
View return policy
Description
Complement Component C3aR Monoclonal antibody specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)Specifications
Complement Component C3aR | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
HC3aRZ8 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
Human Complement Component C3aR transfectants | |
0.1 mL | |
GPCR | |
719 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction