Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C3aR Antibody (HC3aRZ8), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314618JF646
Description
Complement Component C3aR Monoclonal antibody specifically detects Complement Component C3aR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)Specifications
Complement Component C3aR | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
HC3aRZ8 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
Human Complement Component C3aR transfectants | |
0.1 mL | |
GPCR | |
719 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction