Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C5/C5a Antibody (06), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306182MFV500
Description
Complement Component C5/C5a Monoclonal antibody specifically detects Complement Component C5/C5a in Human samples. It is validated for ELISASpecifications
Complement Component C5/C5a | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
727 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
06 | |
Sandwich ELISA Capture | |
C5a;C5b;C5D;Complement C5;CPAMD4;ECLZB | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Component C5/C5a (Accession#: NP_001726.2; Leu679-Arg751). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction