Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C5a Antibody (442), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289580APC
Description
Complement Component C5a Monoclonal antibody specifically detects Complement Component C5a in Human samples. It is validated for ELISASpecifications
Complement Component C5a | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
442 | |
ELISA | |
C5, C5A, HE | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Complement Component C5a (Accession#: NP_001726.2; Leu679-Arg751). | |
0.1 mL | |
Immunology, Innate Immunity, Stem Cell Markers | |
727 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction