Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C5aR1 Antibody (P12/1), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066539C
Description
Complement Component C5aR1 Monoclonal specifically detects Complement Component C5aR1 in Human, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Complement Component C5aR1 | |
Monoclonal | |
DyLight 650 | |
50 mM sodium borate with 0.05% sodium azide | |
C5AR1 | |
C5aR - peptide: Met1 - Asn31 | |
Primary | |
Recognizes the C5a receptor (C5aR) CD88, which is predominantly expressed on cells of the myeloid lineage. Clone P12/1 was raised against a synthetic peptide comprising the N-terminal extracellular domain of the C5aR (met1-Asn31). Clone P12/1 does not inhibit the binding of C5a to its receptor. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
P12/1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1 | |
Mouse | |
0.1 mL | |
728 | |
Human, Rhesus Macaque | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction