Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C5aR1 Rabbit anti-Human, Clone: 32-G1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP3120850.025MG
Description
Complement Component C5aR1 Monoclonal antibody specifically detects Complement Component C5aR1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Complement Component C5aR1 | |
Monoclonal | |
1 mg/mL | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1 | |
This antibody was raised by immunizing mice a synthetic Complement Component C5aR1 C-terminal phosphopeptide that contained phosphoserine instead of serine at position 334 and also at the neighboring phosphorylation sites Ser-332 and Ser-338. | |
0.025 mg | |
GPCR, Immunology, Innate Immunity | |
728 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Western Blot, Immunofluorescence | |
32-G1 | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction