Learn More
Complement Component C9 Antibody (22), Alexa Fluor™ 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP221612AF350
Catalog No. NP221612A30
Only null left
Add to Cart
Description
Description
Complement Component C9 Monoclonal specifically detects Complement Component C9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Complement Component C9
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
C9
Complement factor C9 adsorbed onto aluminum hydroxide gel.
Primary
Human
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
22
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
complement component 9, complement component C9
Mouse
0.1 mL
735
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.