Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Component C9 Antibody (22), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221612F
Description
Complement Component C9 Monoclonal specifically detects Complement Component C9 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
Complement Component C9 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
C9 | |
Complement factor C9 adsorbed onto aluminum hydroxide gel. | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
22 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence | |
complement component 9, complement component C9 | |
Mouse | |
Protein A purified | |
RUO | |
735 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction