Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Component C9 Antibody (WU13-15), Novus Biologicals™
SDP

Human Monoclonal Antibody

$243.50 - $731.50

Specifications

Antigen ComplementComponentC9
Clone WU13-15
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP262803
SDP
View Documents
Novus Biologicals
NBP262803
100 μg
Each for $731.50
Only null left
Add to Cart
 
NB019777
SDP
View Documents
Novus Biologicals
NBP2628030.025MG
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
Description

Description

Complement Component C9 Monoclonal specifically detects Complement Component C9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.
Specifications

Specifications

ComplementComponentC9
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay
Unconjugated
Human
PBS containing 0.1% bovine serum albumin with 0.02% Sodium Azide
735
Neoepitope on the 61 kDa complement component C9, integrated in the terminal complement complex (TCC).
Primary
WU13-15
Monoclonal
Purified
Human
complement component 9, complement component C9
C9
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.