Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Complement Component C9 Human Recombinant Protein
SDP

Catalog No. NBP25163525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25163525 0.25 mg
NBP25163505 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25163525 Supplier Novus Biologicals™ Supplier No. NBP2516350.25MG

A recombinant protein with a His-tag corresponding to amino acids 22-559 of Human Complement Component C9.

A.A. Sequence: MGSSHHHHHH SSGLVPRGSH MGSQYTTSYD PELTESSGSA SHIDCRMSPW SEWSQCDPCL RQMFRSRSIE VFGQFNGKRC TDAVGDRRQC VPTEPCEDAE DDCGNDFQCS TGRCIKMRLR CNGDNDCGDF SDEDDCESEP RPPCRDRVVE ESELARTAGY GINILGMDPL STPFDNEFYN GLCNRDRDGN TLTYYRRPWN VASLIYETKG EKNFRTEHYE EQIEAFKSII QEKTSNFNAA ISLKFTPTET NKAEQCCEET ASSISLHGKG SFRFSYSKNE TYQLFLSYSS KKEKMFLHVK GEIHLGRFVM RNRDVVLTTT FVDDIKALPT TYEKGEYFAF LETYGTHYSS SGSLGGLYEL IYVLDKASMK RKGVELKDIK RCLGYHLDVS LAFSEISVGA EFNKDDCVKR GEGRAVNITS ENLIDDVVSL IRGGTRKYAF ELKEKLLRGT VIDVTDFVNW ASSINDAPVL ISQKLSPIYN LVPVKMKNAH LKKQNLERAI EDYINEFSVR KCHTCQNGGT VILMDGKCLC ACPFKFEGIA CEISKQKISE GLPALEFPNE K

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 0.4M UREA
Gene ID (Entrez) 735
Molecular Weight (g/mol) TMW: 63.4kDa
Quantity 0.25 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias complement component 9, complement component C9
Common Name Complement Component C9
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.