Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor B Antibody (9B8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$426.00
Specifications
Antigen | Complement Factor B |
---|---|
Clone | 9B8 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP22350801
![]() |
Novus Biologicals
NBP2235080.1ML |
0.1 mL |
Each of 1 for $426.00
|
|
|||||
Description
Complement Factor B Monoclonal specifically detects Complement Factor B in Human samples. It is validated for Western Blot, ELISA.Specifications
Complement Factor B | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
629 | |
Complement factor B purified from human plasma and adsorbed to aluminum hydroxide gel | |
Primary | |
Store at 4C. Do not freeze. |
9B8 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Human | |
B-factor, properdin, BFAHUS4, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B | |
CFB | |
IgG1 κ | |
Protein A or G purified | |
Clone 9B8 is specific for the Bb fragment of human factor B. Antibody binding abolishes the formation of C3bBb complexes. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title