Learn More
Invitrogen™ Complement Factor B Polyclonal Antibody

Description
Complement Factor B Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | Complement Factor B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CFB |
| Gene Accession No. | P00751, P04186 |
| Gene Alias | AHUS4; AI195813; AI255840; alternative-complement pathway C3/C5 convertase; ARMD14; B; BF; B-factor, properdin; BFD; C2; C3 proaccelerator; C3 proactivator; C3/C5 convertase; CFAB; CFB; CFBD; complement component 2 (within H-2S); complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; Da1-24; DADB-122G4.5; Factor B; Fb; FBI12; GBG; glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; H2-Bf; histocompatibility 2, complement component factor B; PBF2; properdin; properdin factor B |
| Gene Symbols | CFB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.