Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106888 Supplier Invitrogen™ Supplier No. PA5106888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Factor B/CFB.

Factor B is a component of the alternative complement pathway. The protein is a single chain polypeptide with a molecular mass of 93 kDa. Factor B is cleaved by factor D into the Ba component (30 kDa) and the Bb (63 kDa). Bb together with C3b forms the alternative pathway C3 convertase C3bBb.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CFB
Gene Accession No. P00751, P04186
Gene Alias AHUS4; AI195813; AI255840; alternative-complement pathway C3/C5 convertase; ARMD14; B; BF; B-factor, properdin; BFD; C2; C3 proaccelerator; C3 proactivator; C3/C5 convertase; CFAB; CFB; CFBD; complement component 2 (within H-2S); complement factor B; Complement factor B Ba fragment; Complement factor B Bb fragment; Da1-24; DADB-122G4.5; Factor B; Fb; FBI12; GBG; glycine-rich beta glycoprotein; glycine-rich beta-glycoprotein; H2-Bf; histocompatibility 2, complement component factor B; PBF2; properdin; properdin factor B
Gene Symbols CFB
Host Species Rabbit
Immunogen A synthesized peptide derived from human CFB(Accession P00751), corresponding to amino acid residues T241-K291.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14962, 294257, 629
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.