Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor D/Adipsin Antibody (D10/4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $426.00
Specifications
Antigen | Complement Factor D/Adipsin |
---|---|
Clone | D10/4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019320
![]() |
Novus Biologicals
NBP2235320.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NBP22353201
![]() |
Novus Biologicals
NBP2235320.1ML |
0.1 mL |
Each for $426.00
|
|
|||||
NBP223532
![]() |
Novus Biologicals
NBP223532 |
0.25 mg | N/A | N/A | N/A | ||||
Description
Complement Factor D/Adipsin Monoclonal specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot, ELISA.Specifications
Complement Factor D/Adipsin | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN | |
CFD | |
IgG2a κ | |
Protein A or G purified | |
Clone D10/4 is specific for complement factor D in human serum and plasma |
D10/4 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Human | |
1675 | |
Human Factor D | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title