Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor D/Adipsin Antibody (I8/1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105121B
Description
Complement Factor D/Adipsin Monoclonal specifically detects Complement Factor D/Adipsin in Human samples. It is validated for Western Blot, ELISA.Specifications
Complement Factor D/Adipsin | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CFD | |
Human factor D. | |
0.1 mL | |
Immunology | |
1675 | |
Human | |
Purified |
Western Blot, ELISA | |
I8/1 | |
Western Blot, ELISA | |
Adipsin, ADNcomplement factor D, C3 convertase activator, complement factor D (adipsin), complement factor D preproprotein, D component of complement (adipsin), DF, EC 3.4.21, EC 3.4.21.46, PFD, Properdin factor DADIPSIN | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Clone I8/1 is specific for complement factor D in human serum and plasma | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction