Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor D Monoclonal Antibody (13)
GREENER_CHOICE

Catalog No. PIMA530279
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530279 Supplier Invitrogen™ Supplier No. MA530279
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Adipsin/Complement Factor D.

Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor D
Applications ELISA
Classification Monoclonal
Clone 13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CFD
Gene Accession No. P00746
Gene Alias 28 kDa adipocyte protein; adipsin; Adn; C3 convertase activator; Cfd; Complement factor D; complement factor D (adipsin); complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Df; EC 3.4.21.46; Endogenous vascular elastase; EVE; PFD; Properdin factor D
Gene Symbols CFD
Host Species Mouse
Immunogen Recombinant Human Adipsin/Complement Factor D protein (Ile26-Ala253).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1675
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.