Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Complement Factor D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586828
Description
Purity is > 95% by SDS-PAGE.
Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.
Specifications
Complement Factor D | |
Polyclonal | |
Unconjugated | |
CFD | |
28 kDa adipocyte protein; adipsin; Adn; C3 convertase activator; Cfd; Complement factor D; complement factor D (adipsin); complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Df; EC 3.4.21.46; Endogenous vascular elastase; EVE; PFD; Properdin factor D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11537, 1675, 54249 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P00746, P03953, P32038 | |
CFD | |
A synthetic peptide corresponding to residues in Human CFD. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction