Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596941
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596941 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596941 Supplier Invitrogen™ Supplier No. PA596941
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILGGREAEAH ARPYMASVQL NGAHLCGGVL VAEQWVLSAA HCLEDAADGK VQVLLGAHSL SQPEPSKRLY DVLRAVPHPD SQPDTIDHDL LLLQLSEKAT LGPAVRPLPW QRVDRDVAPG TLCDVAGWGI VNHAGRRPDS LQHVLLPVLD RATCNRRTHH DGAITERLMC AESNRRDSCK GDSGGPLVCG GVLEGVVTSG SRVCGNRKKP GIYTRVASYA AWIDSVLA; Positive Samples: A-549, 22Rv1, THP-1, Mouse lung.

Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CFD
Gene Accession No. P00746, P03953
Gene Alias 28 kDa adipocyte protein; adipsin; Adn; C3 convertase activator; Cfd; Complement factor D; complement factor D (adipsin); complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Df; EC 3.4.21.46; Endogenous vascular elastase; EVE; PFD; Properdin factor D
Gene Symbols CFD
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 26-253 of human CFD (NP_0019192).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11537, 1675
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.