Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547208
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547208 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547208 Supplier Invitrogen™ Supplier No. PA547208
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse Complement Factor D is observed, and less than 1% cross-reactivity with recombinant human (rh) Complement Factor B and rhComplement Factor I is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor D
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CFD
Gene Accession No. P00746
Gene Alias 28 kDa adipocyte protein; adipsin; Adn; C3 convertase activator; Cfd; Complement factor D; complement factor D (adipsin); complement factor D preproprotein; D component (adipsin) of complement; D component of complement (adipsin); Df; EC 3.4.21.46; Endogenous vascular elastase; EVE; PFD; Properdin factor D
Gene Symbols CFD
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Complement Factor D Ile26-Ala253.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1675
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.