Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor H Antibody [Janelia Fluor™ 669], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF4779JF669
Description
Complement Factor H Polyclonal antibody specifically detects Complement Factor H in Human samples. It is validated for Immunohistochemistry,Western BlotSpecifications
Complement Factor H | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Antigen Affinity-purified | |
RUO | |
3075 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Western Blot | |
Janelia Fluor 669 | |
adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246 | |
Mouse myeloma cell line NS0-derived recombinant human Complement Factor H, Ser860-Arg1231, Accession # P08603 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction