Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor H Monoclonal Antibody (C3), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA550911
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550911 200 μL
FEMA550912 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550911 Supplier Invitrogen™ Supplier No. MA550911
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Factor H is a serum protein acting as an inhibitor in the complement activation system. I activates the C3/C5 convertase by dissociating its subunits and also acts as a cofactor for the enzyme activity of Factor I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor H
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene CFH
Gene Alias adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1
Gene Symbols CFH
Host Species Mouse
Immunogen Recombinant Rat CFH protein, Ala975-Ser1217 (Accession #G3V9R2).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155012
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.