Learn More
Invitrogen™ Complement Factor H Monoclonal Antibody (L20/3), Biotin

Description
GAU 020-03B-01 detects Complement factor H (β1H-globulin) in human samples. GAU 020-03B-01 has been successfully used in ELISA procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Specifications
Specifications
| Antigen | Complement Factor H |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | L20/3 |
| Concentration | 1.1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 15 mM sodium azide; pH 7.4 |
| Gene | CFH |
| Gene Accession No. | P08603 |
| Gene Alias | adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.