Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor H Monoclonal Antibody (L20/3)
GREENER_CHOICE

Catalog No. ABG200302
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABG-200302 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABG-200302 Supplier Invitrogen™ Supplier No. GAU0200302
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GAU 020-03-02 detects factor H in human samples. GAU 020-03-02 has been successfully used in ELISA and Western blot procedures. GAU 018-03-02 can be used as a capture antibody in sandwich ELISA with GAU 020-03-02 as detection antibody. This antibody pair is specific for the ~150 kDa Factor H protein. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Factor H is a serum protein acting as an inhibitor in the complement activation system. I activates the C3/C5 convertase by dissociating its subunits and also acts as a cofactor for the enzyme activity of Factor I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor H
Applications ELISA, Western Blot
Classification Monoclonal
Clone L20/3
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene CFH
Gene Accession No. P08603
Gene Alias adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1
Gene Symbols CFH
Host Species Mouse
Immunogen Human factor H.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3075
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.