Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596080 Supplier Invitrogen™ Supplier No. PA596080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDCNELPPRR NTEILTGSWS DQTYPEGTQA IYKCRPGYRS LGNVIMVCRK GEWVALNPLR KCQKRPCGHP GDTPFGTFTL TGGNVFEYGV KAVYTCNEGY QLLGEINYRE CDTDGWTNDI PICEVVKCLP VTAPENGKIV SSAMEPDREY HFGQAVRFVC NSGYKIEGDE EMHCSDDGFW SKEKPKCVEI SCKSPDVING SPISQKIIYK ENERFQYKCN MGYEYSERGD AVCTESGWRP LPSCEEKSCD NPYIPNGDYS PLRIKHRTGD EITYQCRNGF YPATRGNTAK CTSTGWIPAP RCTLK Positive samples: Human plasma, Mouse liver, Mouse plasma, Rat liver; Cellular location: Secreted.

Factor H is a serum protein acting as an inhibitor in the complement activation system. I activates the C3/C5 convertase by dissociating its subunits and also acts as a cofactor for the enzyme activity of Factor I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor H
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene CFH
Gene Accession No. P08603
Gene Alias adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1
Gene Symbols CFH
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3075
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.