Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor H Recombinant Rabbit Monoclonal Antibody (305)
GREENER_CHOICE

Catalog No. PIMA530287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530287 Supplier Invitrogen™ Supplier No. MA530287
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Complement Factor H/CFH.

Factor H is a serum protein acting as an inhibitor in the complement activation system. I activates the C3/C5 convertase by dissociating its subunits and also acts as a cofactor for the enzyme activity of Factor I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor H
Applications ELISA
Classification Recombinant Monoclonal
Clone 305
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7
Gene CFH
Gene Accession No. P08603
Gene Alias adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1
Gene Symbols CFH
Host Species Rabbit
Immunogen Recombinant Human Complement Factor H/CFH protein (Ser860-Arg1231).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.