Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor H-related 2/CFHR2/CFHL2 Antibody (005), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290262MFV500
Description
Complement Factor H-related 2/CFHR2/CFHL2 Monoclonal antibody specifically detects Complement Factor H-related 2/CFHR2/CFHL2 in Human samples. It is validated for ELISASpecifications
Complement Factor H-related 2/CFHR2/CFHL2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3080 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
005 | |
ELISA | |
CFHL2FHR-2, complement factor H-related 2, factor H-related gene 2, FHR2DDESK59, H factor (complement)-like 3, H factor-like 3, H factor-like protein 2, HFL3complement factor H-related protein 2 | |
Rabbit immunized with purified, recombinant Human Complement Factor H-related 2/CFHR2/CFHL2 (Uniprot#: P36980-1, Met1-Lys270) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction