Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor H-related 4/CFHR4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310545100UL
Description
Complement Factor H-related 4/CFHR4 Polyclonal specifically detects Complement Factor H-related 4/CFHR4 in Human samples. It is validated for Western Blot.Specifications
Complement Factor H-related 4/CFHR4 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
complement factor H-related protein 4, CFHL4, complement factor H-related 4, FHR4, FHR-4 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human Complement Factor H-related 4/CFHR4 (NP_006675). Peptide sequence LWVSCANGQEVKPCDFPEIQHGGLYYKSLRRLYFPAAAGQSYSYYCDQNF | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
10877 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction