Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Complement Factor I Monoclonal Antibody (KT22)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA172517
Description
This antibody can be used as a primary antibody for ELISA (1 μg/mL).
Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the active form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the b-chain (38 kDa). Plasma concentration is 35 μg/mL.
Specifications
Complement Factor I | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P05156 | |
CFI | |
Purified Factor B from human blood. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA | |
KT22 | |
Unconjugated | |
CFI | |
AHUS3; ARMD13; C3B/C4B inactivator; C3BINA; C3b-INA; C3b-inactivator; Cfi; complement component factor i; complement component I; complement control protein factor I; complement factor I; Complement factor I heavy chain; Complement factor I light chain; factor i; FI; IF; KAF; Konglutinogen-activating factor; light chain of factor I | |
Mouse | |
Protein A | |
RUO | |
3426 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction