Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575509 Supplier Invitrogen™ Supplier No. PA575509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the active form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the b-chain (38 kDa). Plasma concentration is 35 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor I
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CFI
Gene Accession No. P05156, Q61129, Q9WUW3
Gene Alias AHUS3; ARMD13; C3B/C4B inactivator; C3BINA; C3b-INA; C3b-inactivator; Cfi; complement component factor i; complement component I; complement control protein factor I; complement factor I; Complement factor I heavy chain; Complement factor I light chain; factor i; FI; IF; KAF; Konglutinogen-activating factor; light chain of factor I
Gene Symbols CFI
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 492-537 of Human Complement Factor I.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12630, 3426, 79126
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.