Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complement Factor I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101539 Supplier Invitrogen™ Supplier No. PA5101539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Factor I/CFI.

Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the active form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the b-chain (38 kDa). Plasma concentration is 35 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Factor I
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CFI
Gene Accession No. P05156
Gene Alias AHUS3; ARMD13; C3B/C4B inactivator; C3BINA; C3b-INA; C3b-inactivator; Cfi; complement component factor i; complement component I; complement control protein factor I; complement factor I; Complement factor I heavy chain; Complement factor I light chain; factor i; FI; IF; KAF; Konglutinogen-activating factor; light chain of factor I
Gene Symbols CFI
Host Species Rabbit
Immunogen A synthesized peptide derived from human CFI(Accession P05156), corresponding to amino acid residues I450-G500.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3426
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.