Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Complement Factor MASP3/MASP1 Antibody (1E2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311240
Description
Complement Factor MASP3/MASP1 Monoclonal specifically detects Complement Factor MASP3/MASP1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunoassay.Specifications
Complement Factor MASP3/MASP1 | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Immunoprecipitation, Immunoassay | |
Complement factor MASP-3, Complement-activating component of Ra-reactive factor, CRARF1, CRARFMAP1, DKFZp686I01199, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.104, EC 3.4.21.42, FLJ26383, mannan-binding lectin serine peptidase 1 (C4/C2 activating component ofRa-reactive factor), mannan-binding lectin serine protease 1, mannan-binding lectin serine protease 1 (C4/C2 activating component ofRa-reactive factor), Mannose-binding lectin-associated serine protease 1, Mannose-binding protein-associated serine protease, MAp44, MASP, MASP-1, MASP3, MGC126283, MGC126284, PRSS5, Ra-reactive factor serine protease p100, RaRF, Serine protease 5 | |
The antibody recognizes the heavy chain common to both MASP-1 and MASP-3. | |
50 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Immunoassay | |
100 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
5648 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction