Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement receptor 1 Mouse anti-Human, Unconjugated, Clone: 9H3, MilliporeSigma™

Catalog No. MABF22622MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22622MI 25 μg
MABF22621MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22622MI Supplier MilliporeSigma Supplier No. MABF226225UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Complement receptor 1 Monoclonal antibody specifically detects Complement receptor 1 in Human samples. It is validated for Dot Blot, Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement receptor 1
Applications Dot Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 9H3
Conjugate Unconjugated
Gene Accession No. NP_000564.2
Gene Alias C3b/C4b receptor;CD35
Gene Symbols CR1, C3BR
Host Species Mouse
Immunogen Soluble human complement receptor 1
Molecular Weight of Antigen 223.66 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 9H3 is a mouse monoclonal antibody that detects human complement receptor type 1. It targets an epitope with in the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.