Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement receptor C5aR (CD88) Mouse anti-Human, Clone: S5/1, MilliporeSigma™

Catalog No. MABF198025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1980-25 25 μL
MABF1980-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1980-25 Supplier MilliporeSigma Supplier No. MABF198025UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Complementreceptor C5aR (CD88), clone S5/1 Flow Cytometry, Immunoprecipitation, Inhibition/Activity assays, andn Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement receptor C5aR (CD88)
Applications ELISA, Flow Cytometry, Immunoprecipitation, Inhibition Assays, Western Blot
Classification Monoclonal
Clone S5/1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols C5AR1,C5AR,C5R1
Host Species Mouse
Immunogen BSA-conjugated linear peptide corresponding to the first 31 amino acids from the N-terminal region of human C5a anaphylatoxin chemotactic receptor 1.
Purification Method Protein G Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001727.1
Test Specificity Clone S5/1 specifically detects C5a anaphylatoxin chemotactic receptor 1 (C5aR). It targets an epitope within the first 31 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.