Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Complement Receptor C5aR Rat anti-Mouse, MilliporeSigma™

Catalog No. MABF197625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1976-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1976-25 Supplier MilliporeSigma Supplier No. MABF197625UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This rat monoclonal Anti-Complement Receptor C5aR, clone 20/70 Neutralizing, Activity Assay, Immunocytochemistry and Inhibits Activity/Function, for the detection of C5a anaphylatoxin chemotactic receptor 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Receptor C5aR
Applications Flow Cytometry, Neutralization, Immunocytochemistry, Inhibition Assays
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols C5ar1,C5ar,C5r1
Host Species Rat
Immunogen Rat basophilic leukemia RBL-2H3 cells stably expressing murine C5aR.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 20/70 detects complement 5a receptor in murine species. It targets an epitope with in the extracellular domain.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.