Learn More
Agilent BioTek Complete Direct Drain Waste Systems
Catalog No. BT1170560 Shop All Agilent Technologies Inc. Products
Change view
Click to view available options
Type:
Complete Direct Drain Waste System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
BT1170560 Complete Direct Drain Waste System
BT1170562 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BT1170560 Supplier Agilent BioTek Supplier No. 1170560
This item is not returnable. View return policy
Drains into sink instead of bottles
  • Compatible with Agilent BioTek 405 and 406 Series Microplate Washer Systems
  • Eliminates the need of attending to and emptying waste bottles
  • Saves space on the workbench
TRUSTED_SUSTAINABILITY
Specifications
Includes Direct drain module (115V/230V), standard or high-flow vacuum pump, tubing, holder for two 4L bottles
Type Complete Direct Drain Waste System
For Use With (Equipment) Agilent BioTek 405 Series Microplate Washer System, Agilent BioTek 406 Series Microplate Washer System