Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COX3 Monoclonal Antibody (DA5BC4)
GREENER_CHOICE

Catalog No. 459300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-930-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-930-0 Supplier Invitrogen™ Supplier No. 459300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Isolated mitochondria from Human , Rat and Mouse hearts. WB Predicted molecular weight: 30 kDa.

COX3 is a multi-pass membrane protein. It belongs to the cytochrome c oxidase subunit 3 family. Defects in COX3 are a cause of Leber hereditary optic neuropathy (LHON) and cytochrome c oxidase deficiency (COX deficiency). Defects in MT-CO3 are also found in mitochondrial encephalomyopathy with lactic acidosis and stroke-like episodes (MELAS) syndrome and recurrent myoglobinuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX3
Applications Western Blot
Classification Monoclonal
Clone DA5BC4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene COX3
Gene Accession No. P00414, P00416, P00420, P05505
Gene Alias ACI60_gp01; Co3; Coiii; COX3; COXIII; cytchrome c oxidase sububnit 3; Cytochrome c oxidase polypeptide III; Cytochrome c oxidase subunit 3; cytochrome c oxidase subunit III; mitochondrially encoded cytochrome c oxidase III; Mtco3; mt-Co3; OXI2; Q0275
Gene Symbols COX3, MT-CO3
Host Species Mouse
Immunogen Full length native Yeast Cytochrome C Oxidase subunit III.
Purification Method IgG fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139542, 17710, 24573142, 26204, 4514, 854627
Target Species Human, Mouse, Rat, Yeast
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.