Learn More
Complex Nutrients in Seawater, Whole Volume, WP Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111004657 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
500 mL
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
If a solution is not to be analyzed within two days after opening, preserve a sample aliquot with 0.2 mL of concentrated sulfuric acid per 100 mL of sample (pH<2) and store at 4 degrees C until just before analysis. Analyze the preserved sample as soon as possible within the maximum 28-day holding time.Sample preparationThe sample is ready for immediate analysis.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Kjeldahl nitrogen, total (TKN) 1.5-35 mg/L; Nitrogen, total 1.5-35 mg/L; Phosphorus, total 0.5-10 mg/L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.