Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119011
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119011 200 μL
PIPA5122073 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119011 Supplier Invitrogen™ Supplier No. PA5119011
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COMT Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

Catechol-O-methyltransferase catalyzes the transfer of a methyl group from S-adenosylmethionine to catecholamines, including the neurotransmitters dopamine, epinephrine, and norepinephrine. This O-methylation results in one of the major degradative pathways of the catecholamine transmitters. In addition to its role in the metabolism of endogenous substances, COMT is important in the metabolism of catechol drugs used in the treatment of hypertension, asthma, and Parkinson disease. COMT is found in two forms in tissues, a soluble form (S-COMT) and a membrane-bound form (MB-COMT). The differences between S-COMT and MB-COMT reside within the N-termini. Several transcript variants are formed through the use of alternative translation initiation sites and promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Comt
Gene Accession No. P21964
Gene Alias Catechol O-methyltransferase; catecholamine-O-methyltransferase; catechol-O-methyltransferase; catechol-O-methyltransferase 1; catechol-O-methyltransferase isoform; Comt; Comt1; D16Wsu103e; D330014B15Rik; epididymis secretory sperm binding protein Li 98n; HEL-S-98n; OTTHUMP00000197749; OTTHUMP00000197750; testicular tissue protein Li 42
Gene Symbols Comt
Host Species Rabbit
Immunogen Recombinant protein Catechol-O-Methyltransferase. The antigen corresponds to amino acid range 52-271 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.