Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COMT Polyclonal Antibody, Invitrogen™

Catalog No. PIPA588877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588877 Supplier Thermo Scientific Supplier No. PA588877

Rabbit Polyclonal Antibody

Immunogen sequence: VGDKKGKIVD AVIQEHQPSV LLELGAYCGY SAVRMARLLS PGARLITIEI NPDCAAITQR MVDFAGVKDK VTLVVGASQD IIPQLKKKYD VDTLDMVFLD HWKDRYLPDT LLLEECGLLR KGTVLLADNV ICPGAPDFLA HVRGSSCFEC THYQSFLEYR EVVDGLEKAI YKGPGSEAGP; Positive Samples: MCF-7, HT-29, THP-1, A-431, HepG2, Mouse liver; Cellular Location: Cell membrane, Cytoplasm, Extracellular side, Single-pass type II membrane protein

Catechol-O-methyltransferase catalyzes the transfer of a methyl group from S-adenosylmethionine to catecholamines, including the neurotransmitters dopamine, epinephrine, and norepinephrine. This O-methylation results in one of the major degradative pathways of the catecholamine transmitters. In addition to its role in the metabolism of endogenous substances, COMT is important in the metabolism of catechol drugs used in the treatment of hypertension, asthma, and Parkinson disease. COMT is found in two forms in tissues, a soluble form (S-COMT) and a membrane-bound form (MB-COMT). The differences between S-COMT and MB-COMT reside within the N-termini. Several transcript variants are formed through the use of alternative translation initiation sites and promoters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMT
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene Comt
Gene Accession No. O88587, P21964
Gene Alias Catechol O-methyltransferase, catecholamine-O-methyltransferase, catechol-O-methyltransferase, catechol-O-methyltransferase 1, catechol-O-methyltransferase isoform, Comt, Comt1, D16Wsu103e, D330014B15Rik, epididymis secretory sperm binding protein Li 98n, HEL-S-98n, OTTHUMP00000197749, OTTHUMP00000197750, testicular tissue protein Li 42
Gene Symbols Comt
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 42-221 of human COMT (NP_0092941)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12846, 1312
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.