Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COMTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540116 Supplier Invitrogen™ Supplier No. PA540116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, and HepG2 cells. Predicted reactivity: Mouse (87%), Rat (88%), Rhesus Monkey (98%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

COMTD1 (Catechol O Methyltransferase Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includeO methyltransferase activity. An important paralog of this gene isENSG00000284844.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COMTD1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% Thimerosal; pH 7
Gene COMTD1
Gene Accession No. Q86VU5
Gene Alias 1810030M08Rik; Catechol O-methyltransferase domain-containing protein 1; catechol-O-methyltransferase domain containing 1; catechol-O-methyltransferase domain-containing protein 1; Comtd1; MT773; O-methyltransferase-containing protein; UNQ766/PRO1558
Gene Symbols COMTD1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human COMTD1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118881
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.