Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Concanamycin A, Tocris Bioscience™
SDP

Catalog No. 265610U Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-561-0U 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-561-0U Supplier R&D Systems Supplier No. 2656/10U

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

H+-ATPase (vacuolar) inhibitor

Concanamycin A is a specific inhibitor of V-type (vacuolar) H+-ATPase that displays > 2000-fold selectivity over other H+-ATPases (IC50 values are 9.2, > 20000, > 20000 and > 20000 nM for yeast V-type, F-type, P-type H+-ATPases and porcine P-type Na+,K+-ATPase respectively). Blocks cell surface expression of virus envelope glycoproteins without affecting synthesis and exhibits cytotoxicity in several cell lines.

Chemical Identifiers

CAS 80890-47-7
Molecular Formula C46H75NO14
Molecular Weight (g/mol) 866.10
MDL Number MFCD00210037
InChI Key DJZCTUVALDDONK-HQMSUKCRSA-N
Synonym (3Z,5E,7R,8R,9S,10S,11R,13E,15E,17S,18R)-18-[(1S,2R,3S)-3-[(2R,4R,5S,6R)-4-[[4-O-(Aminocarbonyl)-2,6-dideoxy-β-D-arabino-hexopyranosyl]oxy]tetrahydro-2-hydroxy-5-methyl-6-(1E)-1-propenyl-2H-pyran-2-yl]-2-hydroxy-1-methylbutyl]-9-ethyl-8,
PubChem CID 131859947
IUPAC Name (2R,3S,4R,6R)-6-{[(2R,4R,5S,6R)-2-[(2S,3R,4S)-4-[(2R,3S,4E,6E,9R,10S,11S,12R,13R,14E,16Z)-11-ethyl-10,12-dihydroxy-3,17-dimethoxy-7,9,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6,14,16-tetraen-2-yl]-3-hydroxypentan-2-yl]-2-hydroxy-5-methyl-6-[(1E)-prop-1-en-1-yl]oxan-4-yl]oxy}-4-hydroxy-2-methyloxan-3-yl carbamate
SMILES CC[C@H]1[C@@H](O)[C@H](C)C\C(C)=C\C=C\[C@H](OC)[C@H](OC(=O)\C(OC)=C\C(\C)=C\[C@@H](C)[C@H]1O)[C@@H](C)[C@@H](O)[C@H](C)[C@@]1(O)C[C@@H](O[C@H]2C[C@@H](O)[C@H](OC(N)=O)[C@@H](C)O2)[C@H](C)[C@H](O1)\C=C\C

Specifications

Type Concanamycin A
Quantity 10 μg
Formula Weight 866.09
Percent Purity >90%
Chemical Name or Material Concanamycin A
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.