Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Conditioning Reagent, for Sulfate Analysis (Turbidimetric), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Conditioning Reagent |
Color | Colorless |
CAS | 7732-18-5, 67-63-0, 7647-14-5, 56-81-5, 7647-01-0 |
CAS Min % | 53.0, 13.87, 13.33, 11.22, 6.29 |
CAS Max % | 55.0, 14.44, 13.88, 11.68, 6.54 |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction