Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C 5400 ppm TDS as NaCl, NIST Traceable, Reagents

Catalog No. p-200021354
Click to view available options
Quantity:
500 mL
1 L
4 L

Catalog No. 01-336-656

Supplier: Reagents Holdings Llc C70100001A

Only null left
Add to Cart
Add to Cart

Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C 5400 ppm TDS as NaCl, NIST Traceable, Standard, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C 5400 ppm TDS as NaCl, NIST Traceable
Packaging Poly Bottle
CAS 7732-18-5, 7647-14-5
Specific Gravity 1.0007
Physical Form Liquid
Molecular Formula NaCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 6.7 to 7.3
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Type Conductivity Standard
Quantity 1 L
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.